Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC5B Monoclonal Antibody (5B19-2E)
GREENER_CHOICE

Catalog No. 377400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-740-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-740-0 Supplier Invitrogen™ Supplier No. 377400
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Both IHC and WB applications require the following deglycosylation protocols. Immunohistochemistry 1. Prepare the following stock solutions: a. 200 mM NaIO4 (8.556 g/200 mL water) b. Sodium-Acetate buffer, pH 4.5: Dissolve NaCl (final concentration: 0.33M) into 0.1M glacial acetic acid and adjust to pH 4.5 using 1 N NaOH. c. Sodium-thiosulfate solution: Dissolve Na2S2O3 (0.133M), NaI (0.033M), and NaHCO3 (0.033M) in water and adjust to pH 7.6 using 1M HCl. Keep solution at 4°C. 2. After deparaffinization, rehydration, and peroxidase quenching steps, mix a 1:1 solution of a and b (in step 1), add to each slide, and incubate overnight at 4°C. 3. Treat tissue with solution c for 30 minutes at 4°C. 4. Proceed with normal immunostaining protocol (blocking and antibody incubation steps). Western blotting 1. Treat transblotted membrane with 10 mM NaIO4 in 0.1M sodium acetate/5 mM EDTA, pH 5.5 overnight at 4°C in the dark. 2. After washing off the solution, incubate the membrane in a solution containing 30 mM Na2S2O3, 7.5 mM NaI, and 7.5 mM NaHCO3, pH 7.2 for 1 hour at 4°C in the dark. 3. Proceed with normal Western blot protocol (wash and blocking steps).

Mucins (MUCs) are heavily glycosylated glycoproteins that are synthesized either as membrane-bound or secreted proteins. Mucins are noteworthy because they have an aberrant expression profile in various malignancies. Mucins normally play a protective role for epithelial tissues and are expressed in the gastrointestinal, respiratory, and genito-urinary tracts. Mucins may also be involved in the renewal and differentiation of the epithelium, and the modulation of cell adhesion and cell signaling. MUC5B is thought to play a vital role in maintaining oral health, and may also be involved in respiratory and oral disease.2 MUC5B expression has been studied in gastric carcinoma as well as chronic obstructive pulmonary disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC5B
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5B19-2E
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene MUC5B
Gene Accession No. Q9HC84
Gene Alias Cervical mucin; cervical mucin MUC5B; high molecular weight salivary mucin MG1; MG1; MUC5; MUC5B; MUC-5B; MUC9; mucin 5, subtype B, tracheobronchial; mucin 5B, oligomeric mucus/gel-forming; Mucin-5 subtype B, tracheobronchial; Mucin-5B; sublingual gland mucin
Gene Symbols MUC5B
Host Species Mouse
Immunogen Synthetic peptide derived from the N-terminal region of the human MUC5B protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727897
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.