Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC8 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501736181 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6181 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6181 Supplier Proteintech Group Inc Supplier No. 554891AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to MUC8.

Mucin glycoproteins (Mucins) are major constituents of the glycocalyx that covers mucosal epithelium. MUC8 is one of the major mucins in the ethmoid mucosa and is upregulated by chronic inflammation. TNFa, IL-1b and a combination of both can significantly increase Mucin 8 mRNA levels, suggesting that a mixture of inflammatory mediators can synergistically increase secretion of mucin in human nasal epithelium.

Specifications

Antigen MUC8
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MUC8
Gene Alias MUC8, mucin 8
Gene Symbols MUC8
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100129528
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.