Learn More
Abcam Mucic Acid, Dicarboxylic Acid Tetrol, Powder, 50 g
Description
- Molecular weight: 210.14 g/mol
- Form: Powder
- Composition: 100% Mucic acid
- Description: Dicarboxylic acid tetrol compound formed from the oxidation of galactose
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at ambient temperature. |
| Form | Powder |
| Buffer | 100% Mucic acid |
| Formulation | 100% Mucic acid |
| Quantity | 50 g |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.