Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucobromic acid, 99+%

Catalog No. p-3779254
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128051000 100 g Glass bottle
AC128055000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC128051000 Supplier Thermo Scientific Chemicals Supplier No. 128051000
Only null left
Add to Cart

CAS: 488-11-9 | C4H2Br2O3 | 257.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-11-9
Molecular Formula C4H2Br2O3
Molecular Weight (g/mol) 257.86
MDL Number MFCD00063745
InChI Key NCNYEGJDGNOYJX-IHWYPQMZSA-M
Synonym mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate
PubChem CID 6994897
IUPAC Name (Z)-2,3-dibromo-4-oxobut-2-enoate
SMILES C(=O)C(=C(C(=O)[O-])Br)Br

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 122°C to 125°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula BrC(CHO)=C(Br)CO2H
Quantity 100 g
Beilstein 03, IV, 1721
Formula Weight 257.86
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Mucobromic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-670-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.