Learn More
Thermo Scientific™ Mueller Hinton Agar
Catalog No. R454086 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R454086 10 kg
R454082 500 g
R454084 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R454086 Supplier Thermo Scientific™ Supplier No. R454086
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
TRUSTED_SUSTAINABILITY
Specifications
Description Mueller Hinton Agar
Form Powder
Media Type Mueller Hinton Agar
Product Type Nutrient Agar
Quantity 10 kg