Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mueller Hinton Agar
GSA_VA

Catalog No. R04052
Change view
Click to view available options
Quantity:
10/Pk.
100/Cs.
40/Pk.
Format:
15 x 100mm Plate
15 x 150mm Plate
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R04052 40/Pk. 15 x 150mm Plate
R01620 10/Pk. 15 x 100mm Plate
R01624 100/Cs. 15 x 100mm Plate
R04050 10/Pk. 15 x 150mm Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R04052 Supplier Thermo Scientific™ Supplier No. R04052
Add to Cart
Add to Cart
This item is not returnable. View return policy

Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.

Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar, a medium designed and manufactured according to the European standard, EUCAST, and the American standard, CLSI.

The addition of starch to Mueller Hinton Agar ensures that toxic factors found during growth will be absorbed, and its presence is often essential to establish growth from very small inocula. Conforms to EUCAST, CLSI and DIN.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery We recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF.
Description Mueller Hinton Agar
Format 15 x 150mm Plate
Manufacturing Origin United States
Product Type Agar Plate
Quantity 40/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.