Learn More
Teknova Mueller Hinton Broth for Francisella. 500mL, Sterile. 8 pack.
SDP
Supplier:  Teknova M58898PK
Encompass_Preferred
1.75% Casamino Acids, 0.20% Beef Extract, 0.15% Starch, 0.50% Sodium Chloride, 1.00% Peptone, 0.10% Cerelose Anhydrous Dextrose (Glucose), 0.025% Ferric Pyrophosphate, 2.00% IsoVitalex, 2.50% Calf Serum
Catalog No. 50-238-7488
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.