Learn More
Liofilchem™ Mueller Hinton Fastidious Agar
SDP
Catalog No. 21403471 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
10 Plates
20 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-403-471 10 Plates
21-403-466 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-471 Supplier Liofilchem™ Supplier No. 11132
Add to Cart
Add to Cart
A medium recommended by EUCAST for antimicrobial susceptibility testing of Streptococcus spp, Haemophilus spp and several other fastidious organisms
  • Mueller Hinton Fastidious Agar (Horse blood 5 %, 20 mg/L β-NAD) is recommended by EUCAST for antimicrobial susceptibility testing of Streptococcus spp (including S. pneumoniae), Haemophilus spp and several other fastidious organisms.
  • Meat extract and casamino acids supply amino acids and other nitrogenous substances, minerals, vitamins, carbon and other nutrients to support the growth of microorganisms.
  • Starch acts as a protective colloid against toxic substances that may be present in the medium, and hydrolysis of starch during autoclaving provides a small amount of glucose which is a source of energy.
  • Agar is the solidifying agent.
  • Horse blood and β-NAD are a further growth supplementation for fastidious microorganisms.
Specifications
Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class Fastidious, Haemophilus species
Test Type Antimicrobial Susceptibility
Quantity 10 Plates
Diameter (Metric) 140 mm