Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liofilchem™ Mueller Hinton II Agar + 2% NaCl
SDP

Catalog No. 21403472 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-403-472 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-403-472 Supplier Liofilchem™ Supplier No. 11206
Add to Cart
Add to Cart

A medium for susceptibility testing of methicillin resistant staphylococcus aureus (MRSA)

  • Mueller Hinton II Agar + 2% NaCl is specifically used for testing the antibiotic susceptibility of Staphylococcus spp., particularly for detecting methicillin resistance (MRSA/MRSE) via disc diffusion or gradient test, because the high salt concentration (2% NaCl) promotes the expression of resistance genes (mecA), giving accurate results for these salt-tolerant bacteria.
  • It is standardized by CLSI/EUCAST for this purpose, ensuring reliable detection of resistance phenotypes like methicillin-resistant S. aureus (MRSA). 2% NaCl creates a hypertonic environment, forcing Staphylococcus to express the resistance mechanism, leading to smaller inhibition zones and correctly identifying resistant isolates.
  • This medium is manufactured to contain low levels of thymine and thymidine and controlled levels of calcium and magnesium ions.

Specifications

Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class S. aureus
Test Type Antimicrobial Susceptibility
Quantity 20 Plates
Diameter (Metric) 90 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.