Learn More
Liofilchem™ Mueller Hinton II Agar
SDP
Catalog No. 21403463 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
10 Plates
20 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-403-463 20 Plates
21-403-467 10 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-463 Supplier Liofilchem™ Supplier No. 10031
Add to Cart
Add to Cart
A medium for antimicrobial susceptibility testing by the disc diffusion technique, according to CLSI and EUCAST
  • With increasing antimicrobial resistance, AST of aerobic bacteria becomes more relevant. Mueller Hinton II Agar permits excellent growth of most aerobic bacteria.
  • This medium is formulated to have low levels of thymine and thymidine and is adjusted to the calcium and magnesium ions concentrations.
  • It conforms to Clinical and Laboratory Standards Institute (CLSI) and European Committee on Antimicrobial Susceptibility Testing (EUCAST) recommendations.
  • Its formulation, with controlled levels of ions (Ca++, Mg++) and low thymine/thymidine, ensures reliable antibiotic diffusion and bacterial growth for accurate susceptibility results.
Specifications
Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class Aerobic Microorganisms, Gram-negative Organisms, Gram-positive Organisms, Multidrug-resistant, Gram-negative Organisms, Bacteria
Test Type Antimicrobial Susceptibility, Microbial Susceptibility, Antibiotic Susceptibility Sensitivity, Minimum Inhibitory Concentration (MIC), Microbial Cultivation
Quantity 20 Plates
Diameter (Metric) 90 mm