Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liofilchem™ Mueller Hinton II Agar (Sheep Blood 5%)
SDP

Catalog No. 21403465 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-403-465 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-403-465 Supplier Liofilchem™ Supplier No. 10131
Add to Cart
Add to Cart

A medium for antimicrobial susceptibility testing of certain fastidious organisms by the disc diffusion technique, as standardized by the CLSI

  • Mueller Hinton II Agar (Sheep Blood 5%) is a medium recommended for antimicrobial susceptibility testing of Streptococcus spp. and Neisseria meningitidis by the disc diffusion technique (Kirby-Bauer method), as standardized by the Clinical and Laboratory Standard Institute (CLSI).
  • Mueller Hinton II Agar (Sheep Blood 5%) is manufactured to contain low levels of thymine and thymidine and controlled levels of calcium and magnesium ions.
  • The sheep blood provides essential nutrients for these less robust bacteria, which would not grow well on unsupplemented Mueller Hinton Agar.

Specifications

Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class Fastidious
Test Type Antimicrobial Susceptibility
Quantity 20 Plates
Diameter (Metric) 90 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.