Learn More
Azzota Corp CERAMIC FIBER MUFFLE FURNACE, 1200℃, 2L, 1500W, 110V, LCD display, Inner Chamber size (W*L*H): 120*200*80mm
Supplier:  Azzota Corp SX4212PB
Encompass_Preferred
CERAMIC FIBER MUFFLE FURNACE, 1200℃, 2L, 1500W, 110V, LCD display, Inner Chamber size (W*L*H): 120*200*80mm
Catalog No. NC2753694