Learn More
Abcam MUG, Fluorescent Beta-Glucuronidase Substrate, >99% Purity, 250 mg
Description
- Molecular weight: 388.33 g/mol
- Purity: >99%
- Description: Fluorescent substrate for β-glucuronidase that yields a blue fluorescent end product upon cleavage, detectable by UV light.
- Applications: Typically used in the β-glucuronidase (GUS) reporter system for detecting gene expression in Escherichia coli and transformed plants.
- Chemical composition: 100% (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxane-2-carboxylic acid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | β-Glucuronidase |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | MUG |
| Product Type | Liquid Substrate |
| Reactivity | Escherichia coli, Plant |
| Detection Method | Fluorescence |
| Content And Storage | Store at -20°C. |
| Cross Reactivity | Escherichia coli, Plant |
| Quantity | 250 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.