Learn More
V&P Scientific Inc MULTI-BLOT REPLICATOR, 96 Wells, Standard Plate, Flat Tip, Delivers ~200nL, 1.58mm Pin Diameter
Supplier:  V&P Scientific Inc VP 408
Encompass
MULTI-BLOT REPLICATOR, 96 Wells, Standard Well Plate, Stainless Steel, Flat Tip, Delivers ~200nL Hanging Drop, 1.58mm Pin Diameter, 22mm Total Length, 9mm Centers
Catalog No. NC2160429
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.