Learn More
Description
- Gel-style hand cleaner clings to hands and spreads easy.
- Contains natural pumice scrubbers.
- USDA BioPreferred™ Certified.
- EcoLogo™ Certified.
- Readily biodegradeable.
- Contains a natural citrus solvent.
- Meets California VOC requirement for consumer products.
- Effective on oil, grease, paint, tar, and asphalt.
Specifications
Specifications
| Product Type | Heavy Duty Hand Cleaner |
| Color | Green |
| For Use With (Application) | Oil, Grease, Oil Based Paint, Tar, Asphalt, Mud, Clay, Soil, Brake Dust, Carbon Black, Shellacs, Urethanes, Varnish, Wood Stain, Gasket Sealers, Asphalt Sealers, Creosote, Modern Printing Ink |
| Quantity | 4 Pk. |
| Volume (English) | 0.5 gal. |
| Volume (Metric) | 1.89 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
