Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Multi-kinase inhibitor 1 | 778274-97-8 | MFCD29035151 | 99.1% | 418.37 g/mol | C20H17F3N4O3 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1030325MG

Encompass_Preferred

Multi-kinase inhibitor 1 is a small-molecule research reagent that inhibits multiple tyrosine kinases, including PDGF-R, c-Kit, and Bcr-Abl. It is supplied as an off-white to yellow solid and is used in biochemical and cellular studies to probe receptor tyrosine kinase signaling.

  • Potent inhibition of PDGF-R, c-Kit, and Bcr-Abl.
  • High purity (99.05%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Provided with datasheet, SDS, and COA for characterization.
  • Molecular formula C20H17F3N4O3; molecular weight 418.37 g/mol.
  • Suitable for biochemical and cellular assay applications.

Catalog No. 50-002-63796


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.