Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ Multi-Layer Isolation Gown

Catalog No. 22415812
This item is not returnable. View return policy
Catalog No. 22-415-812 Supplier Dukal™ Supplier No. 315GSS
This item is not returnable. View return policy

SMS multi-layer isolation gown features elastic wrists with waist and neck tie closures.

  • Made of quality multi-layer material
  • Features elastic wrists with waist tie and neck closures

Specifications

Product Type Isolation Gown
For Use With (Application) Personal Protection
Size Universal
Gender Unisex
No. of Pockets None
Material SMS
Color Yellow
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Autoclavable Not Autoclavable
Show More Show Less

Not made with natural rubber latex

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.