Learn More
Alkali Scientific Clear Immunoplate PS 96 Well P
SDP
Supplier:  Alkali Scientific MPRUNI
Encompass_Preferred
ASI96-Well Solid Plates Clear polystyrene surface chemistry for optimal binding is hydrophilic and ideal for protein research and antibody sandwich assaysAlphanumerically labeled rows and columns Multibinding Suitable specifically for adsorbing hydrophilic molecules (Antigen ELISA) Maxibinding Modified polystyrene surface for higher binding capacity for proteins and other molecules with both hydrophilic and hydrophobic regions (Recommended for most ELISA experiments) Medibinding Surface adsorbs proteins and molecules that consists of interrmediate hydrophobic and hydrophilic properties (Antigen/Antibody ELISA More hydrophbic compared to Maxibinding Plates) Unibinding Suitable for adsorption of hydrophobic molecules (Antigen ELISA) Plate type (96well per plate) Lid not included 3 type bottom Flat / Round / Flat-Small Developed for immunoassay(ELISA) and general bin
Catalog No. 50-302-8493
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.