Learn More
Glas Col LLC Vortexer
SDP
Supplier:  Glas Col LLC 099A VB4
Encompass_Preferred
Exclusive pulsing feature interrupts vortex to join lower and upper strata. Interchangeable cartridges eliminate the need for extra racks.Two cartridges included, one for 12-13mm and one for 15-16mm diameter tubes. 100-2000 RPM, 120V, 1.5A, 50/60 Hz.
Catalog No. NC9912344
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.