Learn More
Description
- Product Benefits:
- Low-angle bevel design allows easy penetration
- Color-coded caps and hubs allow easy identification of needle gauge size
Specifications
Specifications
| Product Type | Multi-sample Needle |
| Sterility | Sterile |
| Disposable | Single-use |
| For Use With (Equipment) | Fisherbrand Blood Collection Tube Holder |
| Gauge | 22 G |
| Latex Free | Yes |
| Length (English) | 1 in. |
| Preattached Holder | No |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.