Learn More
Thermco Products Multi-Trip USB Humidity and Temperature Data Logger
Catalog No. 04500044
Change view
Click to view available options
Display Type:
LCD
Catalog No. Display Type
04-500-044 LCD
1 options
Catalog No. 04-500-044 Supplier Thermco Products Supplier No. LTUHADO16
Add to Cart
Add to Cart
Automatically generates a PDF report without special software. Audible alarm and red LED signals a temperature alarm.
  • The LogTag™ temperature and humidity logger records up to 16,000 high-resolution readings.
  • Measures humidity from 0 to 100% RH and temperature from -30°C to +70°C (-22°F to +158°F).
  • Onboard display shows current temperature, humidity, alarm status, and recording status.
  • Review button allows viewing of minimum and maximum temperature and humidity values for the trip, along with the duration of any alarm-range events.
  • Configuration and data download are supported via a micro USB to USB cable, with an option to auto-generate PDF reports.
  • Equipped with three contact pins for compatibility with LogTag™ LTI-HID and LTI-WiFi Interface Cradles.
  • Data can be processed using LogTag Analyzer software or the LogTag Online cloud platform for comprehensive analysis and storage.
Specifications
Type Temperature and Humidity Datalogger
Display Type LCD
Alarm Types Audible, LED Indicator