Learn More
United Scientific™ Multi-Use Bench Pulley
SDPSCI_ED
Catalog No. S16036 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type: Table Mount Pulley
Catalog No.
S16036 Product Type: Table Mount Pulley
1 options
Catalog No. S16036 Supplier United Scientific™ Supplier No. PULB01
Add to Cart
Add to Cart
Ideal for use in demonstrating simple machines.
  • High quality bench pulley that can be mounted on a support stand rod.
  • Constructed of a heavy duty cast aluminum support frame, this 50 mm diameter pulley is mounted on heavy duty low friction cone bearings.
  • Fits table tops up to 1.75 in. thick (4.5 cm).
Specifications
Product Type Table Mount Pulley
Warning: Cancer and reproductive harm