Learn More
Cytiva (Formerly Pall Lab) Multi-well Plate Vacuum Manifold
Catalog No. NC9079007 Shop All Cytiva Products
Encompass
Designed to Perfectly Fit AcroPrep™, AcroPrep Advance, and AcroWell™ Filter Plates-Comes complete with the necessary O-ring and gasket. The control block includes the vacuum pressure gauge, vacuum metering valve, vacuum release valve, and 14 in. hose barb for vacuum line attachment.-Vacuum manifold unit includes a Delrins spacer block designed to accommodate standard 350 L receiver plates. The spacer block has been optimized to reduce the space between the receiver plate and the filter plate during vacuum filtration.-Optional spacer block available for use with 1 mL receiver plates.-Adapter collar holds filter plates tightly to receiver plates for centrifugation. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC9079007 Supplier Cytiva Supplier No. 5017
Designed to Perfectly Fit AcroPrep™, AcroPrep Advance, and AcroWell™ Filter Plates-Comes complete with the necessary O-ring and gasket. The control block includes the vacuum pressure gauge, vacuum metering valve, vacuum release valve, and 14 in. hose barb for vacuum line attachment.-Vacuum manifold unit includes a Delrins spacer block designed to accommodate standard 350 L receiver plates. The spacer block has been optimized to reduce the space between the receiver plate and the filter plate during vacuum filtration.-Optional spacer block available for use with 1 mL receiver plates.-Adapter collar holds filter plates tightly to receiver plates for centrifugation. This product is not approved for use in medical, clinical, surgical or other patient applications.