Learn More
Anton Paar Multicube CT is the control unit for Multicube 48
Supplier:  Anton Paar 196910
Encompass_Preferred
Multicube CT’s capacitive touchscreen can be calibrated to reliably respond even when operated with gloves. Easily adjust the temperature in steps of 0.1 °C at any time via the touchscreen. Choose between heating ramp, heating as fast as possible, or definition of a stepwise ramp of up to 10 temperature steps to reach the target temperature. The temperature and time is shown on the touchscreen throughout the process. Store up to 100 processing methods (incl. implemented standard methods and Anton Paar’s recommended methods). Suitable for use with Anton Paar Multicube 48 Hot block digestion heating unit NC1818247
Catalog No. NC1818251
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.