Learn More
Sigma Aldrich Fine Chemicals Biosciences AscentisR Express F5 2.7 m
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 53569U
Encompass
The pentafluorophenylpropyl stationary phase of Ascentis Express F5 provides a stable reversed phase packing with electron-deficient phenyl rings due to the presence of electronegative fluorines.In addition to forming pi-pi and mildly steric interactions F5 phases also retain compounds by polar interactions. Ascentis Express F5 can be used for basic acidic or neutral compounds with alternate selectivity from C18.
Catalog No. 50-290-8963
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.