Learn More
AdipoGen MultimericCD40L hAnim-Free
SDP
Supplier:  AdipoGen AG40B0010AFC100
Encompass_Preferred
Recombinant protein. Human CD40L aa 116-261 is fused at the N-terminus to mouse ACRP30headless aa 18-111 and a FLAG -tag. Source CHO cells. Produced using animal component-free medium. Lyophilized. Contains PBS Induces B cell activation as demonstrated by dose-dependent upregulation of CD86 ED50 <1ng/ml. B cell expansion online protocol available. CD40 ligand CD40L expressed by activated T cells engages CD40 expressed by B cells and APCs. CD40L can provide signals to B cells that induce proliferation, immunoglobulin class switching and antibody secretion. CD40L has been used recently in Immunotherapy to activate tumor-infiltrated lymphocytes TIL. MultimericCD40L™ is a high-activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo. It provides a simple and equally potent alternative to combinations. Mul
Catalog No. 50-256-2938
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.