Learn More
Chemglass Life Sciences Multiple Rod Connector
SDP
Supplier:  Chemglass Life Sciences CG926801
Encompass_Preferred
Multiple Rod Connector. Total adjustment capability within one connector. Permits lattice adjusting after construction. Holds rods at 90° while allowing horizontal and vertical adjustment. Side and front holes accommodate a variety of configurations. Accepts rod size 1/2” (13mm).
Catalog No. CG926801