Learn More
New England Biolabs, Inc. NEBNext Multiplex Oligos for Illumina (Index Primers Set 2) – 24 reactions
SDP
Supplier:  New England Biolabs, Inc. E7500S
Encompass
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
Catalog No. NC0509778
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.