Learn More
Cellular Engineering Technologies Multipotent Unrestricted Somatic Stem Cell Expansion Media (450mL)
Supplier:  Cellular Engineering Technologies MR1006
Encompass_Preferred
Multipotent unrestricted somatic expansion media is designed to support the robust growth of cord blood stem cells. Media has been shown to maintain stem cells undifferentiated for up to 10 passages. Media is compatible with available animal-origin serum, supplemented separately according to the customer preference.Media is shipped with gel packs.
Catalog No. NC2753068
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.