Learn More
Axygen™ MultiRack Racked Tips
Catalog No. 07202397 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
300 μL
1250 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Extended Length
Extended Length, Low Retention
Extended Length, Low Retention, Maxymum Recovery
Universal
Universal, Low Retention
Rack Color:
Blue
Green
Red
Yellow
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Tip Style Rack Color
07-202-397 1250 μL Non-sterile Extended Length Blue
07-202-393 10 μL Non-sterile Extended Length Red
07-202-398 10 μL Sterile Extended Length Red
07-202-403 10 μL Sterile Extended Length, Low Retention Red
07-202-394 200 μL Non-sterile Universal Yellow
07-202-399 200 μL Sterile Universal Yellow
07-202-404 200 μL Sterile Universal, Low Retention Yellow
07-202-395 300 μL Non-sterile Universal Green
07-202-400 300 μL Sterile Universal Green
07-202-401 300 μL Sterile Extended Length, Low Retention Green
07-202-405 300 μL Sterile Universal, Low Retention Green
07-202-406 300 μL Sterile Extended Length, Low Retention, Maxymum Recovery Green
07-202-402 1250 μL Sterile Extended Length Blue
07-202-407 1250 μL Sterile Extended Length, Low Retention Blue
07-202-396 300 μL Non-sterile Extended Length Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 07-202-397 Supplier Axygen™ Supplier No. MR1000XTR
Add to Cart
Add to Cart
This item is not returnable. View return policy
MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.
The Axygen MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter, and reload packaging styles to meet the variety of needs in the lab setting. The MultiRack tip system offers standard and extended length tips covering volumes from 1 μL to 1250 μL. For racked and reload packaging styles, the tips are pre-loaded into easy-load color-coded deck inserts. With the smaller footprint and reusable rack, this system uses 30% less plastic, minimizing the environmental impact. The stackable design saves critical bench space. Axygen MultiRack Tip System products are made in the USA and meet ISO standards. All tips are free of detectable RNase, DNase, DNA, and pyrogens. For more details visit our Certificates page at www.corning.com/lifesciences.
Features
• Reusable rack with reload systems 
• Smaller footprint and stackable design, saving valuable bench space
• Extended tips offered in 10, 300, and 1250 μL (1000 μL extended length) volumes
• Color-coded racks for easy volume identification
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1250 μL
Filtered Non-filtered
Sterility Non-sterile
Color Blue
Rack Color Blue
Autoclavable Autoclavable
Model MR-1000XT-R
Certifications/Compliance ISO 13845
Clean Claims Free of detectable DNAse/RNAse and human DNA; Non pyogenic
Format Hinged Rack
Tip Style Extended Length
Packaging Hinged rack
Quantity 768/pk, 5 pks/cs
Product Type Pipette Tips
Color Code None
Disposable Single-use
Graduated Yes
Product Line MultiRack
Material Polyethylene
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov