Learn More
Description
- MultiScreen™ 384-well polypropylene microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices.
- Polypropylene microplates are appropriate for sample preparation and storage in addition to some biochemical assays.
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Array | 16 x 24 |
| Color | Natural |
| Product Type | Microplate |
| Material | Polypropylene |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Surface Treatment | Untreated |
| Well Shape | Conical Bottom (V-well) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.