Learn More
Millipore Multiscreen™ 96-Well Plate, Polypropylene, Solid Bottom
Catalog No. MSPNNVX00 Shop All MilliporeSigma Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
MSPNNVX00 Conical Bottom (V-well)
MSPNNFX00 Flat Bottom (F-well)
MSPNNUX00 Round Bottom (U-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MSPNNVX00 Supplier Millipore Supplier No. MSPNNVX00
Only null left
Add to Cart
Add to Cart
Non-sterile polypropylene plates for use in common biochemical assay platforms.
  • MultiScreen™ 96-well polypropylene microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices.
  • Polypropylene microplates are appropriate for sample preparation and storage in addition to some biochemical assays.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Color Natural
Product Type Microplate
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)