Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreenHTS Durapore™ 96-Well Filter Plates

Catalog No. MSBVS1210 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
0.65 μm
1.2 μm
Color:
Clear
Opaque
Material:
Barex
Styrene
Quantity:
10/Pk.
50/Pk.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Color Material Quantity
MSBVS1210 1.2 μm Clear Styrene 10/Pk.
MSBVN1B50 1.2 μm Opaque Barex 50/Pk.
MSBVN1250 1.2 μm Clear Styrene 50/Pk.
MSBVN1210 1.2 μm Clear Styrene 10/Pk.
MSDVN6B50 0.65 μm Opaque Barex 50/Pk.
MSGVN2210 0.22 μm Clear Styrene 10/Pk.
MSGVN2250 0.22 μm Clear Styrene 50/Pk.
MSHVN4B10 0.45 μm Opaque Barex 10/Pk.
MSHVS4510 0.45 μm Clear Styrene 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MSBVS1210 Supplier MilliporeSigma™ Supplier No. MSBVS1210
Add to Cart
Add to Cart

Designed for full automation compatibility. MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.

For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.
  • Standardized dimensions and aligned, rigid sidewalls
    • Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
  • Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
  • Extended skirt and vacuum manifolds
    • Allows easy, contamination-free stacking directly with collection plates
    • Protects recessed underdrain
    • Provides ample surface for bar code labels
    • Improves vacuum seal
  • Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Opaque plastic prevents light transfer between wells during direct scintillation counting
  • Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
  • Meet ANSI™/SBS 2004 plate specifications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SBS
Format Standard
Membrane Hydrophilic PVDF
Array 8 x 12
Pore Size 1.2 μm
Binding Type Low Binding
Color Clear
Material Styrene
No. of Wells 96
Sterility Sterile
Filtration Area 0.28 cm2
For Use With (Application) Bead based assays
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.