Learn More
Description
Designed for full automation compatibility. MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.
- Standardized dimensions and aligned, rigid sidewalls
- Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
- Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
- Extended skirt and vacuum manifolds
- Allows easy, contamination-free stacking directly with collection plates
- Protects recessed underdrain
- Provides ample surface for bar code labels
- Improves vacuum seal
- Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
- Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
- Opaque plastic prevents light transfer between wells during direct scintillation counting
- Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
- Meet ANSI™/SBS 2004 plate specifications
Specifications
Specifications
| Certifications/Compliance | ANSI, SBS |
| Format | Standard |
| Membrane | Hydrophilic PVDF |
| Array | 8 x 12 |
| Pore Size | 0.65 μm |
| Binding Type | Low Binding |
| Color | Opaque |
| Material | Barex |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.