Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MultiShot™ TOP10 Chemically Competent E. coli Cells, 96-well plate

Catalog No. A40007058
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96 Well Plate (20 μL/Well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40007058 1 x 96 Well Plate (20 μL/Well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40007058 Supplier Invitrogen™ Supplier No. A40007058
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

MultiShot TOP10 Chemically Competent E. coli Cells provide high-efficiency transformation for routine cloning in an automation-ready 96-well format. Ideal for stable propagation of high-copy plasmids and blue/white screening in high-throughput workflows.

Invitrogen MultiShot TOP10 Chemically Competent E. coli Cells are derived from DH10B cells and are one of the most popular laboratory strains. Provided in a clear skirted 96-well microtiter plate to simplify high-throughput workflows and provide transformation efficiencies of >1 x 108 cfu/μg control plasmid DNA, Multishot TOP10 cells are a versatile strain that can be used in multiple routine cloning applications.

Features of MultiShot TOP10 competent cells include:

  • TOP10 strain ideal for high-efficiency cloning and plasmid propagation
  • Microtiter plate format design convenient for automated high-throughput cloning
  • mcrA mutation that supports efficient transfection of methylated DNA
  • lacZΔM15 allele that permits rapid blue/white screening of transfectants
  • endA1 mutation that helps reduce endonuclease activity and improve plasmid DNA yields
  • recA1 mutation helps reduce unwanted recombination

MultiShot TOP10 competent cells in 96-well plates are designed to facilitate high-throughput transformation. This format allows all steps of the transformation protocol, up to plating, to take place in the same well, saving time and reducing the chance of contamination. After the addition of DNA, MultiShot TOP10 competent cells can be transformed by heat-shock at 42°C using a heat block or thermocycler in the same plate.

Genotype

F-mcrA Δ(mrr-hsdRMS-mcrBC) Φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara, leu)7697 galU galK rpsL (StrR) endA1 nupG

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Includes pUC19 DNA, S.O.C media
Transformation Efficiency Level ≥1.0 x 108
Shipping Condition Dry Ice
Species E. coli
Format 96-well Plate
Product Line MultiShot
For Use With (Application) Cloning
Quantity 1 x 96 Well Plate (20 μL/Well)

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.