Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MultiShot™ FlexPlate DH10B T1R Competent Cells

Catalog No. C6481201
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96 well plate (20 μl/well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C6481201 1 x 96 well plate (20 μl/well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C6481201 Supplier Invitrogen™ Supplier No. C6481201
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Box 1:

  • MultiShot™ FlexPlate DH10B™ T1R Chemically Competent E. coli
  • One 96-well plate, 20 μl per well

Box 2:

  • pUC19 DNA (10 pg/μl): 1 vial, 50 μl
  • S.O.C. medium: 2 bottles, 10 ml each

MultiShot FlexPlate DH10B T1R chemically competent E. coli cells are cloning competent cells pre-aliquoted in a 96-well PCR plate to increase transformation productivity.

MultiShot FlexPlate DH10B T1R chemically competent E. coli cells are cloning competent cells pre-aliquoted in a 96-well PCR plate to increase transformation productivity. The FlexPlate format can be conveniently divided into 8-well segments for flexibility in transformation throughput.

All of our T1R strains have the tonA (fhuA) deletion in their genotype that confers resistance to T1 and T5 phage. T1 bacteriophage spread rapidly and lyse E. coli hosts, which are commonly used for cloning and library construction. A resistant genotype offers added security to protect valuable clones and libraries. This is especially important for genome and sequencing centers where T1 phage infection would be catastrophic.

Diverse DH10B T1R cloning capabilities
MultiShot FlexPlate DH10B T1R competent cells may be used in a variety of cloning applications as they are resistant to the effects of ligase and ligation buffers and provide transformation efficiencies of >1 x 108 cfu/μg with control plasmid DNA. The cells are ideal for cloning large plasmids and BACs and are suitable for cloning DNA that contains methylcytosine and methyladenine (i.e., genomic DNA).

MultiShot FlexPlate DH10B T1R Competent Cell features
• mcrA, mcrBC, mrr, and hsdRMS elimination of restriction systems to allow construction of more representative genomic constructs
• tonA (fhuA) genotype to confer resistance to T1 and T5 phage
• endA1 for increased plasmid yield and quantity
• recA1 for reduced occurrence of non-specific recombination in cloned DNA
• lacZΔM15 for blue/white color screening of recombinant clones

Increase transformation productivity with MultiShot FlexWell format
The FlexPlate is a 96-well PCR plate that can be used in automated liquid handling systems or divided into 8-well segments for flexibility in transformation throughput. Addition of DNA to the MultiShot FlexPlate Competent Cells can be quickly performed with a multichannel pipet or a liquid handling system. After addition of DNA, the cells can be efficiently transformed by heat-shock using a water bath, heat block, or thermal cycler.

Key benefits of the MultiShot FlexPlate format include:
• Increase throughput—compatible with automated liquid handling platforms
• Flexible 96-well plate may be divided into 8-wellsstrips
• Competent cells come pre-aliquoted in 96-well plates and ready for transformation

Genotype
F- mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 lacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK rpsL nupG λ- tonA

Find the strain and format that you need
• We offer many E. coli strains and formats of chemically competent and electrocompetent cells to meet your specific transformation needs.
• Choose from our MultiShot formatted comp cells for high-throughput options.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Cloning Methylated DNA Yes
Content And Storage • MultiShot FlexPlate DH10B T1R Competent Cells (1 x 96-well plate at 20 μL/well)
Store at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store at –20°C.

• S.O.C. Medium (2 x 10 mL)
Store at 4°C or room temperature.

Antibiotic Resistance Bacterial No
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility High
Plasmid May be used for plasmids >100 kb
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (K12)
Format 96-well Plate
Product Line Multishot
Quantity 1 x 96 well plate (20 μl/well)
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.