Learn More
Beckman Coulter Multisizer™ 4e Smart Technology Aperture Tubes
Smart Technology Aperture Tubes are designed specifically for the Multisizer 4 or 4e Analyzer. The aperture allows extended sizing measurement range of 2% to 80% of the aperture diameter.
Supplier:  Beckman Coulter C93985
 View more versions of this product
Catalog No. 15-655-147
Only null left
Add to Cart
Description
Description
  • Extended measurement range (2% to 80% of aperture diameter) leads to less frequent aperture changes for more analysis options
  • A wide range of apertures is available with diameters ranging from 10 to 2,000 μm
  • Specifically designed for the Multisizer 4e or Multisizer 4 Coulter Counters
  • 100 μm ST Aperture Tube is standard with the Multisizer 4e Coulter Counter
  • Compatible with ST Beakers and Accuvette STs
  • Convenient tubes ensure accurate, and consistent results
  • Size and serial number information is included on a bar code on the tube for automatic data entry via instrument bar code reader
Specifications
Specifications
Aperature Tube
8.0 μm to 320 μm
Multisizer™ 4 or 4e Coulter Counters
Glass
400 μm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.