Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Multiskan SkyHigh Microplate Spectrophotometer

Catalog No. 502068595
Encompass
Encompass_Preferred
Change view
Click to view available options
Interface:
On-board or PC Control
Touchscreen
Cuvettes:
No
Yes
Includes:
μDrop Duo plate
Plate not included
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Interface Cuvettes Includes
50-206-8595 Touchscreen Yes μDrop Duo plate
50-206-8591 On-board or PC Control No Plate not included
50-206-8592 Touchscreen No Plate not included
50-206-8593 Touchscreen No μDrop Duo plate
50-206-8594 Touchscreen Yes Plate not included
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-206-8595 Supplier Thermo Scientific™ Supplier No. A51119700DPC
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

Thermo Scientific Multiskan SkyHigh Microplate Spectrophotometer is an easy to use instrument for photometric and turbidimetric applications, especially for DNA, RNA and protein analyses, including endpoint, kinetic, and spectral assays. Results are shown on Multiskan SkyHigh models with a large high-resolution touch screen. SkanIt Software is a powerful tool to address the challenging applications.

Multiskan SkyHigh Microplate Spectrophotometer is a UV/Vis microplate reader designed to be convenient and easy to use for any photometric absorption or turbidimetric research applications and especially DNA, RNA and protein analysis. This microplate reader is ideal for multi-user environments where a variety of endpoint, kinetic and spectral assays are performed. Multiskan SkyHigh is available in different configurations. Touchscreen models offer the flexibility to use the system as stand-alone, or together with Thermo Scientific™ SkanIt Software for PC control. A model with a cuvette reading option is also available. For convenient data handling, Multiskan SkyHigh has an option to access to Thermo Fisher Connect and Microsoft OneDrive cloud-based tools.

  • Monochromator-based optical system for free selection of wavelengths from 200 nm to 1000 nm
  • Compatible with 6-48, 96- and 384-well microplates or cuvettes with maximum plate height of 19.5 mm including possible lid
  • Reads μDrop™ and μDrop Duo Plates for micro-volume DNA/RNA and protein analysis
  • Separate optimized measurement modes for absorption and turbidimetric measurements
  • Allows kinetic, spectral, and endpoint measurements for a variety of applications
  • Fast operation: full spectrum of a well is done in less than 10 seconds; typical DNA spectrum can be measured in less than three seconds and a full 96-well microplate is read with one wavelength in 6 seconds
  • Performs onboard shaking and incubation for temperature-critical or cell-based assays
  • Touchscreen models have an easy-to-use interface for stand-alone use, and includes ready-made protocols for UV based nucleic acid and protein quantification as well as colorimetric protein quantification
  • Thermo Fisher Connect or Microsoft OneDrive cloud-based tools allow you to securely store, access, share, and manage your photometric data remotely (touchscreen models)
  • Multiple languages available; choose between English, German, French, Spanish, Italian, Portuguese, Russian, Chinese, and Japanese (touchscreen models)
  • Setting up of more demanding assays is easy via the powerful and intuitive SkanIt™ Software
  • Multi-instrument software platform compatible with all Thermo Scientific microplate readers
  • Open license software for unlimited installation to multiple computers
  • Research Edition for life sciences; Drug Discovery Edition with features for compliance with FDA 21 CFR Part 11
  • User-interface languages available: English, French, German, Spanish, Portuguese, Italian, Russian, Japanese, and Chinese
  • Searchable SkanIt Cloud Library with ready-to-use protocols including calculations and true application example data
  • Measurement sessions are easy to setup and navigate
  • Intuitive Virtual Pipetting Tool assists in defining the plate layout
TRUSTED_SUSTAINABILITY

Specifications

Cuvettes Yes
Interface Touchscreen
Incubator Temperature Ambient +2° to 45°C
Light Source Xenon Flash Lamp
Measurement Speed 96 wells: 6 sec., 384 wells: 10 sec.
Plate Type 6, 48, 96, and 384-well Plates, μDrop Plates
Product Line Multiskan
Reading Options Top
Shaking Linear
Spectral Scanning Speed 10 sec. from 200 to 1000 nm
Wavelength Selection Monochromator
Software Type SkanIt Software, Open License Software, Multi-Instrument Software
Accuracy (Photometric) 1.0% + 0.003 Abs (0 to 2.0 Abs), 2.0% (2.0 to 2.5 Abs) at 450 nm
Bandwidth <2.5 nm
Depth (English) 17.5 in.
Depth (Metric) 44.5 cm
Height (English) 10.4 in.
Height (Metric) 26.5 cm
Includes μDrop Duo plate
Linearity 0 to 2.5 Abs, 2% at 450 nm
Resolution 1 nm
Software Language Versions English, German, French, Spanish, Italian, Portuguese, Russian, Chinese, Japanese
Wavelength Range 200 to 1000 nm
Weight (English) 23.8 lb.
Weight (Metric) 10.8 kg
Width (English) 11.6 in.
Width (Metric) 29.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.