Learn More
Siemens Healthineers Multistix™ and Uristix™ Reagent Strips
GSA_VA
Catalog No. AM2164 Shop All Siemens Healthineers Products
Change view
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity
Glucose, Leukocyte, Nitrite, Protein
Description:
Multistix 10SG
Multistix 5
Multistix 7
Multistix 8 SG
Uristix 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Description
AM-2164 Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity Multistix 8 SG
23-042305 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen Multistix 10SG
23-042-351 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen Multistix 5
AM-2166 Glucose, Leukocyte, Nitrite, Protein Uristix 4
23-042310 Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein Multistix 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. AM-2164 Supplier Siemens Healthineers Supplier No. 10337913
Add to Cart
Add to Cart
This item is not returnable. View return policy
Visually and instrumentally readable urinalysis strips
  • Includes testing for a variety of parameters
  • Supplied in lightweight, unbreakable, plastic vials
  • Provide diagnostically useful information about the status of carbohydrate metabolism, kidney and liver function, acid-base balance and bacteriuria-pyuria
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Multistix 8 SG
Detectable Analytes Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity
For Use With (Application) Urinalysis
Quantity 100 Pack
WARNING: Reproductive Harm - www.P65Warnings.ca.gov