Learn More
Anton Paar MULTIWAVE 3001 60 HZ
Supplier:  Anton Paar 324970
Encompass_Preferred
  • Microwave digestion system for efficient sample preparation.
  • Maximum sample capacity of 41 samples per run.
  • 10.1 in high-resolution touchscreen user interface.
  • ETL and GS safety certifications.
  • Over 600 pre-installed digestion programs.
  • SmartVent technology for overpressure release in vessels.
  • Tool-free handling of vessels and rotors.
  • State-of-the-art cooling system.
  • Automated data export ensures traceability.
  • Compliant with pharmacopeia, GMP, GAMP 5, and 21 CFR Part 11 regulations.
  • Operates at a frequency of 60 Hz.
Catalog No. NC3597481
Only null left
Add to Cart
Request a Quote