Learn More
LICOR Multiwell Plate Alignment Guide (guide for 1 plate)
Supplier:  LICOR 92672000
Encompass_Preferred
The 6 Multiwell Plate Alignment Guide is suitable for aligning up to 6 multiwell plates on the glass surface of the Odyssey DLx Imager. The 1 Multiwell Plate Alignment Guide is suitable for aligning one multiwell plate on the glass surface of the Odyssey M or Odyssey DLx Imager.
Catalog No. NC2225474