Learn More
LI COR BIOTECH LLC Multiwell Plate Alignment Guide (guide for 6 plates)
Supplier:  LI COR BIOTECH LLC 92672200
Encompass_Preferred
The 6 Multiwell Plate Alignment Guide is suitable for aligning up to 6 multiwell plates on the glass surface of the Odyssey DLx Imager. The 1 Multiwell Plate Alignment Guide is suitable for aligning one multiwell plate on the glass surface of the Odyssey M or Odyssey DLx Imager.
Catalog No. NC2225475
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.