Learn More
Genscript Corporation MuLV Titer p30 ELISA Kit
Supplier:  Genscript Corporation L01041
Encompass_Preferred
GenScript s MuLV Titer p30 ELISA Kit (Cat No L01041) is a Sandwich ELISA designed for quantitatively measuring the physical titer of Murine leukemia viruses
Catalog No. 50-000-58046
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.