Learn More
Description
PA1-742 detects Munc-18 from human, rat and mouse tissues. PA1-742 has been successfully used in Western blot, immunohistochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects an ~67 kDa protein representing Munc-18 from rat and mouse whole brain extract. PA1-742 immunizing peptide corresponds to amino acid residues 58-70 from human Munc-18. This sequence is completely conserved between human, mouse, rat, bovine, and canine Munc-18. PA1-742 immunizing peptide (Cat. # PEP-067) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | MUNC18 |
| Applications | Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | Stxbp1 |
| Gene Accession No. | O08599, P61764, P61765 |
| Gene Alias | AI317162; AI326233; ANC18HA; EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; MMS10-G; Ms10g; MUNC18-1; Munc-18a; munc-18c; neuronal SEC1; NSEC1; n-sec1; NSEC1A; p67; Protein unc-18 homolog 1; protein unc-18 homolog A; rbSec1; Rb-sec1; rbSec1A; rbSec1B; rop; RP11-56D16.3; Sec1; Stxbp1; Sxtbp1; syntaxin binding protein 1; syntaxin-binding protein 1; UNC18; UNC-18 homolog; unc-18-1; Unc18-1; UNC18A; unc-18A; Unc18h |
| Gene Symbols | Stxbp1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
