Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connecticut Clean Room Corp. Munising LTD Cleanroom Paper

Catalog No. 19807710
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-807-710 Blue
19-807-712 Green
19-807-711 Pink
19-807-714 White
19-807-713 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-807-710 Supplier Connecticut Clean Room Corp. Supplier No. PB2S002LTD
Add to Cart
Add to Cart

High grade, economical cleanroom paper that contains less synthetic polymers and opacity

  • Economical alternative to Munising™ LP
  • Less synthetic polymers, used to seal individual fibers into sheet
  • No natural latex
  • Autoclavable
  • Free of inorganic fillers, minimizes inadvertent doping of products
  • Accepts standard printing inks without skipping, smudging or smearing
  • Heat resistant for use in laser printers and photocopy machines

Specifications

Color Blue
Dimensions (L x W) 8.5 x 11 in.
Autoclavable Autoclavable
For Use With (Application) Cleanrooms
Quantity 2500 Sheets/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.