Learn More
Connecticut Clean Room Corp. Munising LTD Cleanroom Paper
Description
- Economical alternative to Munising™ LP
- Less synthetic polymers, used to seal individual fibers into sheet
- No natural latex
- Autoclavable
- Free of inorganic fillers, minimizes inadvertent doping of products
- Accepts standard printing inks without skipping, smudging or smearing
- Heat resistant for use in laser printers and photocopy machines
Specifications
Specifications
| Color | Pink |
| Dimensions (L x W) | 8.5 x 11 in. |
| Autoclavable | Autoclavable |
| For Use With (Application) | Cleanrooms |
| Quantity | 2500 Sheets/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.