Learn More
Clontech Labs 3P Mupid-exU (Direct Only)
SDP
Supplier:  Clontech Labs 3P AD140
Encompass
The Mupid-exU System gel electrophoresis equipment has several novel features including multichannel pipettor compatibility, a UV-transmissible tank, cover placement light sensor, 7 output voltage settings (18V, 25V, 35V, 50V, 70V, 100V and 135V) and a timer function. These added features enhance the Mupid-exU System's usability and safety.
Catalog No. NC1456007
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.