Learn More
Abcam Mupirocin, Antibiotic Agent, Solid, 100 mg
Description
- Form: Solid
- Molecular Weight: 500.62 g/mol
- Storage: Store at Ambient
- Shipping: Ambient - Can Ship with Ice
- Composition: 100% Mupirocin
- CAS Number: 12650-69-0
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Mupirocin |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Form | Solid |
| Buffer | 100% mupirocin |
| Content And Storage | Store at ambient temperature. |
| Formulation | 100% Mupirocin |
| Quantity | 100 mg |
| For Use With (Application) | Antibiotic research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.