Learn More
Abcam Mupirocin, Antibiotic Agent, Solid, 20 mg
Description
- CAS Number: 12650-69-0
- Form: Solid
- Molecular Weight: 500.62 g/mol
- Constituents: 100% Mupirocin
- Storage Temperature: Store at Ambient
- Shipping Temperature: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Mupirocin |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% Mupirocin |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Formulation | 100% Mupirocin |
| Quantity | 20 mg |
| For Use With (Application) | Antibiotic agent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.