Learn More
Description
- To be performed by agar dilution only
Specifications
Specifications
| Format | Paper Disc |
| Product Type | Antimicrobial Susceptibility Disc |
| Target Organism Class | Gram-positive Microorganisms |
| Test Type | Antimicrobial Susceptibility |
| Species | Staphylococcus aureus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
